Hand cleanser dispensers, Eurospender, 1 Plus Signal
Supplier: BODE CHEMIE
Certificates
About this item
These Eurodispenser 1 plus signal dispensers in high-visible colour are designed to improve hand-hygiene compliance. Colours draw users' attention and may improve compliance with hand disinfection.
- Easy pump exchange through removal from the front, easy bottle replacement
- Reliable robust metal dispenser, easy-to-use, not autoclavable
- Adjustable dosing amount of 0,75 to 1,5 ml per actuation
Frequently Bought Together