Order Entry
France
ContactUsLinkComponent
Volumetric flasks for sugar analysis, class A
Volumetric flasks for sugar analysis, class A
Catalog # ROGO307055
Supplier:  ROGO SAMPAIC
undefined
Volumetric flasks for sugar analysis, class A
Catalog # ROGO307055
Supplier:  ROGO SAMPAIC
Specifications
  • Capacity:
    50/55 ml
  • Height:
    155 mm
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3.3, class A.
  • With two graduation marks
  • Good chemical resistance
  • Good temperature resistance