Order Entry
France
ContactUsLinkComponent
Disinfectant, concentrate, Bomix® plus
  BODE980321
undefined
Disinfectant, concentrate, Bomix® plus
  BODE980321
  • Capacity:
    5 L
  • Description:
    Bomix® plus, aldehyde-free disinfectant
  • Seq:
    03
  • Packed:
    Canister
  • Pk:
    5 L
Bomix® plus is used for manual reprocessing of heat-sensitive and heat-resistant instruments in immersion baths and for the coarse and brush cleaning of endoscopes. Also suitable for flexible endoscopes heavily contaminated with protein.
  • Contents in 100 g: N,N-Didecyl-N-methyl-poly(oxyethyl)ammoniumpropionate 17,5 g and other ingredients like surfactants, solvents, complexing agents, corrosion inhibitors, foam regulators, pH regulators, colourants and fragrances
  • Bactericidal, fungicidal, virucidal against enveloped viruses, polyomavirus and rotavirus
  • pH value for 0,3%, 1%, 2% of solution: approx 7
  • Suitable for ultrasonic baths