Order Entry
France
ContactUsLinkComponent
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates
undefined
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99 %
  • Ignition residue:
    Max. 0.005 %
  • Water:
    Max. 2 %
  • Cl (Chloride):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Ca (Calcium) (0.1 %):
    Max. 0.2 ppm
  • K (Potassium) (0.1 %):
    Max. 0.2 ppm
  • Mg (Magnesium) (0.1 %):
    Max. 0.2 ppm
  • Na (Sodium) (0.1 %):
    Max. 0.5 ppm
  • Transmittance (215 nm) (0.1 %):
    Min. 10 %
  • Transmittance (225 nm) (0.1 %):
    Min. 50 %
  • Transmittance (230 nm) (0.1 %):
    Min. 80 %
  • Transmittance (240 nm) (0.1 %):
    Min. 95 %
  • Transmittance (245 nm) (0.1 %):
    Min. 99 %
  • Suitable for LC-MS (0.1 %):
    Passes test

Specification Test Results

Frequently Bought Together