Tubing, FEP
: METROHM
FEP, translucent.
- With light and kink protection
- With thread connections at both ends
Material allows much longer lengths and tighter tolerances than PTFE tubing.
Frequently Bought Together
FEP, translucent.
Material allows much longer lengths and tighter tolerances than PTFE tubing.