Double bossheads
Supplier: BOCHEM
Certificates
About this item
Double bossheads, either made of aluminium or malleable cast iron, chromium plated.
- For rods up to 16,5 mm
- Angle: 90°
- With safety screw type 'heart' (M8)
Certifications: DIN 12895
Frequently Bought Together