Storage Bags
Supplier: SEMADENI
Jump to:
About this item
Storage bags in transparent LDPE, with bottom weld.
They are also suitable for liquids but without side weld.
Packaging: Minimum quantity:100 pieces
10 Options Available Below
Product Details & Documents
Storage bags in transparent LDPE, with bottom weld.
They are also suitable for liquids but without side weld.
Packaging: Minimum quantity:100 pieces
Product Family Options
Product Information
- ThicknessDimensionsAvailabilityPrice
- SEMA2439Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,05 mm80×130 mmLog in for availability1 * 100 Pce4,20 €
- SEMA2440Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,05 mm100×150 mmLog in for availability1 * 100 Pce11,50 €
- SEMA2442Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,05 mm100×200 mmLog in for availability1 * 100 Pce11,50 €
- SEMA2446Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,05 mm180×300 mmLog in for availability1 * 100 Pce23,30 €
- SEMA2447Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,05 mm200×370 mmLog in for availability1 * 100 Pce30,40 €
- SEMA2448Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,04 mm240×380 mmLog in for availability1 * 100 Pce28,30 €
- SEMA2450Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,075 mm260×400 mmLog in for availability1 * 100 Pce47,40 €
- SEMA2452Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,04 mm300×500 mmLog in for availability1 * 100 Pce39,00 €
- SEMA2455Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,075 mm400×650 mmLog in for availability1 * 100 Pce100,00 €
- SEMA4447Storage bags, transparent, with ground weld.Storage bags, transparent, with ground weld.0,05 mm900×900 mmLog in for availability1 * 100 Pce147,00 €