About this item
TubeFiller was designed to reduce repetitive manual filling of any format laboratory racked tubes. Based on its dedicated function to fill 'any tubes in any rack' the design includes a very stable filling arm with two side rails for best position recovery, precise XY position axes, an easy manual height adjustment, and an integrated pump to cover the standard microbiology filling range. No need for an additional pump for standard applications. Under the tubes there is only stainless steel. Easy to clean, no rims or gaps and therefore microbiology friendly and optimal for food, cosmetic or pharma laboratory environments. Suitable for any rack from small 96-well format, typical microbiology racks holding high glass tubes, up to customised beaker/bottle holders or any other regular racked laboratory containers can become filled. The software allows simplest set-up of XY oriented tube racks. The racks are stored in a library. Any new rack set-up of the filling platform can be programmed via the rack drag & drop menu. The integrated laser tracking allows definition of exact rack position by checking the first and the last tube in the rack. The variable rack frame allows the exact repositioning of rack’s. For repetitive routine applications customer design frames guarantee exact rack positions and process optimised high throughput filling applications. Filling Speed DosiPump DP1000: 2400 ml/min
- Free positioning of any rack
- Intuitive software
- Integrated pump
- True walk away tube/bottle filling
- 2100 cm² free usable surface
- High tubes/bottles up to 25 cm