Capillary columns, traditional phases, SPB-1, Supelco®
: Merck
These non-polar methyl silicone bonded poly(dimethyl siloxane) columns are used to separate the sample components according to boiling point.
- Bonded polymer matches the polarity of its nonbonded predecessors
- Temperature limit from −60 to 320 °C
: Meets USP G1, G2 and G9 requirements.
Frequently Bought Together