Dialysis tubing, SERVAPOR®
Supplier: Serva
Certificates
About this item
Dialysis tubing is made of cellulose membrane. It contains low amounts of impurities such as sulphides and heavy metals.
- High chemical and physical resistance
- pH range: 2 to 12
- Temperature range: 4 to 60 °C
Frequently Bought Together