Milk testing pipettes
Catalog # DUMALLC011
Supplier: DUMAS
Specifications
- Capacity:11 ml
- Description:Milk testing pipette, glass, capillary end
- Pk:1
Specifications
About this item
These milk testing pipettes with capillary tip are made of glass.