Order Entry
France
ContactUsLinkComponent
L(+)-Tartaric acid 25 g/l in aqueous solution
L(+)-Tartaric acid 25 g/l in aqueous solution
Catalog # PRLSMC307199.2000
Supplier:  PROLABO CMP
CAS Number:  
undefined
L(+)-Tartaric acid 25 g/l in aqueous solution
Catalog # PRLSMC307199.2000
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

About this item

 :
 :150.09 g/mol
Storage Temperature:Ambient
EINECS:200-066-2
CAS Number:87-69-4
Formula:C₄H₆O₆
MDL Number:MFCD00064207