L(+)-Ascorbic acid 80 g/l in aqueous solution
Plastic bottle, 5 L
Catalog # PRLS85737.360
Supplier: PROLABO CMP
L(+)-Ascorbic acid 80 g/l in aqueous solution
Catalog # PRLS85737.360
Supplier: PROLABO CMP
CAS Number: 50-81-7
Jump to:
Product Details & Documents
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Storage Temperature:Ambient
- EINECS:200-066-2
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:5 L



