Order Entry
France
ContactUsLinkComponent
Ammonium heptamolybdate 14 g/l in aqueous solution
Ammonium heptamolybdate 14 g/l in aqueous solution
Catalog # PRLS87411.400
Supplier:  PROLABO CMP
CAS Number:  
Ammonium heptamolybdate 14 g/l in aqueous solution
Catalog # PRLS87411.400
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214
  • Environmentally Preferable:

Specifications

About this item

 :
 :1235.86 g/mol
EINECS:234-722-4
CAS Number:12054-85-2
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059