Order Entry
France
ContactUsLinkComponent
Cerium(IV) sulphate 0.4 mol/l (0.4 N) in aqueous solution
Cerium(IV) sulphate 0.4 mol/l (0.4 N) in aqueous solution
Catalog # PRLS89304.5120
Supplier:  PROLABO CMP
CAS Number:  
Cerium(IV) sulphate 0.4 mol/l (0.4 N) in aqueous solution
Catalog # PRLS89304.5120
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate
 :
 :332.24 g/mol
Merck Index:13,02002
EINECS:237-029-5
UN:3264
ADR:8,III
 :GHS07
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852