Isonicotinamidine hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6345-27-3
Formula:C₆H₇N₃·ClH
MDL Number:MFCD00040385
:157.601 g/mol
Melting Pt:∼ 245 °C (dec.)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together