Picolinamidine hydrochloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51285-26-8
:
Formula:C₆H₈ClN₃
MDL Number:MFCD00052271
:157.60 g/mol
Melting Pt:148…152 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together