Chromatography tubing, FEP
Supplier: VICI JOUR RESEARCH
Certificates
About this item
FEP, natural or coloured.
- Solid colours identify the tubing Int.Ø.
- Recommend for ion chromatography applications
FEP (FluorinatedEthylenePropylene) tubing is chemically inert to most HPLC solvents, and is recommended for low pressure applications.
Frequently Bought Together