Order Entry
France
ContactUsLinkComponent
HPLC columns, Hypersil™ BDS Phenyl
HPLC columns, Hypersil™ BDS Phenyl
Certificates
undefined
HPLC columns, Hypersil™ BDS Phenyl
Specifications
  • Catalog No:
  • 555-5952
  • 555-5936
  • 555-5910
  • 555-5909
  • 555-5908
  • Carbon load:
  • 5%
  • 5%
  • 5%
  • 5%
  • 5%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Surface area:
  • 170 m²/g
  • 170 m²/g
  • 170 m²/g
  • 170 m²/g
  • 170 m²/g
  • Temperature limit:
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • USP code:
  • L11
  • L11
  • L11
  • L11
  • L11