Saffron alcoholic solution, Q Path® for microscopy
Pouch, 450 mL
Supplier: VWR Chemicals
Saffron alcoholic solution, Q Path® for microscopy
Catalog # 10047028.
Supplier: VWR Chemicals
CAS Number: 84604-17-1
Jump to:
Product Details & Documents
Ready to use Saffon solution for HES histological staining in easy to use and safe pouch.
Synonyms: C.I. 75100
- CAS Number:84604-17-1
- Molecular Weight:976.96 g/mol
- Storage Temperature:Ambient
- UN:1170
- ADR:3,II
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Colour of solution:Orange
- Density (20/4):0,8 - 0,81
- pH (20°C):7,2 - 7,6
- Absorbance (0.5 %):440 - 443 nm
Specifications
- Pack type:Pouch
- Pk:450 mL
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



