Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Paraffin
Paraffin, liquid, sterile
Paraffin, liquid, sterile
Paraffin, liquid, sterile
Catalog # 301440ZK
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Paraffin, liquid, sterile
Catalog # 301440ZK
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pk:
100 mL
Seq:
2440
Specifications
About this item
Product Details
Documentation
:
CAS Number:
8012-95-1
:
64742-51-4
Product Details
Documentation