4-Methyl-2-(tributylstannyl)pyridine 96%
Catalog # H51472.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:301652-23-3
:
Formula:C₁₈H₃₃NSn
MDL Number:MFCD07787403
:382.16 g/mol
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient