Tubing sleeves, FEP, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
FEP, natural or coloured.
FEP tubing sleeves are an excellent choice for lower pressure applications. These 1,6" long sleeves facilitate connecting capillary tubing into ports designed for 1/32" Ext.Ø. tubing. A wide selection of appropriate sleeve for capillary Ext.Ø. size. These sleeves can be used at up to 50 °C and have a pressure rating of 1750 psi (121 bar).
Frequently Bought Together