Single channel pipettes, electronic, variable volume, E4™ XLS™
About this item
The E4™ XLS™ is an electronic pipette that can be upgraded and expanded right from the PC. Available as standard versions or with LTS LiteTouch System for lower ejection forces.
- Large screen, joystick control and carousel menus speed access to all modes and options
- True Manual™ pipetting mode permits continuous Real Time control of pipetting speed and direction, combined with precise volume measurement
- Independent control of aspirate, dispense and mix speeds for challenging liquids
- Blowout and automatic dispensing options for precision pipetting protocols
- Ergonomic fingerhook and joystick provide maximum comfort
Standard functions include basic pipetting, multi-dispensing, and mixing. Advanced functions include sequential volume programming, fixed volume, titration, dilution and reverse mode pipetting.
Delivery information: Supplied with Li-Ion battery and wall power supply. An optional charging stand is available separately.
17 Options Available Below
Product Details & Documents
The E4™ XLS™ is an electronic pipette that can be upgraded and expanded right from the PC. Available as standard versions or with LTS LiteTouch System for lower ejection forces.
- Large screen, joystick control and carousel menus speed access to all modes and options
- True Manual™ pipetting mode permits continuous Real Time control of pipetting speed and direction, combined with precise volume measurement
- Independent control of aspirate, dispense and mix speeds for challenging liquids
- Blowout and automatic dispensing options for precision pipetting protocols
- Ergonomic fingerhook and joystick provide maximum comfort
Standard functions include basic pipetting, multi-dispensing, and mixing. Advanced functions include sequential volume programming, fixed volume, titration, dilution and reverse mode pipetting.
Delivery information: Supplied with Li-Ion battery and wall power supply. An optional charging stand is available separately.
Product Family Options
LTS models
Product Information
- VolumeIncrementAccuracyImprecisionWithAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Traditional models
Product Information
- VolumeIncrementAccuracyImprecisionWithAvailabilityPrice
- 0,5 - 10 µl0,01 µl2,5 - 1,0 %1,2 - 0,4 %Traditional conical shaftLog in for availability1 * 1 Pce1 490,00 €Specifications:MarketsourceMarketsource
- 10 - 100 µl0,1 µl3,5 - 0,8 %1,0 - 0,15 %Traditional conical shaftLog in for availability1 * 1 Pce1 480,00 €Specifications:MarketsourceMarketsource
- 2 - 20 µl0,02 µl7,5 - 1,0 %2,0 - 0,3 %Traditional conical shaftLog in for availability1 * 1 Pce1 430,00 €Specifications:MarketsourceMarketsource
- 20 - 200 µl0,2 µl2,5 - 0,8 %1,0 - 0,15 %Traditional conical shaftLog in for availability1 * 1 Pce1 430,00 €Specifications:MarketsourceMarketsource
- 20 - 300 µl0,2 µl2,5 - 0,4 %1,0 - 0,15 %Traditional conical shaftLog in for availability1 * 1 Pce1 430,00 €Specifications:MarketsourceMarketsource
- 100 - 1000 µl1,0 µl3,0 - 0,8 %0,6 - 0,15 %Traditional conical shaftLog in for availability1 * 1 Pce1 430,00 €Specifications:MarketsourceMarketsource
- 200 - 2000 µl2,0 µl3,0 - 0,8 %0,6 - 0,12 %Traditional conical shaftLog in for availability1 * 1 Pce1 480,00 €Specifications:MarketsourceMarketsource
- 500 - 5000 µl5,0 µl2,4 - 0,6 %0,6 - 0,16 %Traditional conical shaftLog in for availability1 * 1 Pce1 490,00 €Specifications:MarketsourceMarketsource
- 1000 - 10000 µl10,0 µl5,0 - 0,6 %0,6 - 0,16 %Traditional conical shaftLog in for availability1 * 1 Pce1 490,00 €Specifications:MarketsourceMarketsource
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- SmartStand SCS-B