0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Sodium hydroxide
Sodium hydroxide 0.0326 mol/l (0.0326 N) in aqueous solution
Sodium hydroxide 0.0326 mol/l (0.0326 N) in aqueous solution
PRLS88189.290
:
PROLABO CMP
:
Sodium hydroxide 0.0326 mol/l (0.0326 N) in aqueous solution
PRLS88189.290
:
PROLABO CMP
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
40 g/mol
:
1 g/cm³ (25 °C)
:
Ambient
:
215-185-5
:
1824
:
8,III
:
1310-73-2
:
NaOH
:
MFCD00003548