Potassium chloride 0.008 mol/l (0.008 N) in aqueous solution
PRLSMC307658.1000
:
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
: Potassiumchloride
:
:74.55 g/mol
:Ambient
:231-211-8
:7447-40-7
:KCl
:MFCD00011360