0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Sodium hydroxide
Sodium hydroxide 0.03 mol/l (0.03 N) in methanol
Sodium hydroxide 0.03 mol/l (0.03 N) in methanol
PRLSMC302567.0500
:
PROLABO CMP
:
Sodium hydroxide 0.03 mol/l (0.03 N) in methanol
PRLSMC302567.0500
:
PROLABO CMP
:
Pack type:
Plastic bottle
Pk:
500 mL
Seq:
2725
:
:
40 g/mol
:
Ambient
:
215-185-5
:
3286
:
3,II
:
1310-73-2
:
NaOH
:
MFCD00003548