Order Entry
France
ContactUsLinkComponent
Silver nitrate 25% in aqueous solution
Corrosive
Environment
Silver nitrate 25% in aqueous solution
Catalog # PRLSMC307209.0500
Supplier:  PROLABO CMP
CAS Number:  
undefined
Corrosive
Environment
Silver nitrate 25% in aqueous solution
Catalog # PRLSMC307209.0500
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:1760
ADR:8,I
 :GHS05
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414