Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium thiosulphate
Sodium thiosulphate 0.0001 mol/l (0.0001 N) in aqueous solution
Sodium thiosulphate 0.0001 mol/l (0.0001 N) in aqueous solution
Sodium thiosulphate 0.0001 mol/l (0.0001 N) in aqueous solution
PRLS87463.290
:
PROLABO CMP
:
Sodium thiosulphate 0.0001 mol/l (0.0001 N) in aqueous solution
PRLS87463.290
:
PROLABO CMP
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
158.11 g/mol
:
Ambient
:
231-867-5
:
7772-98-7
:
Na₂S₂O₃
:
MFCD00003499