
Sodium hydroxide 0.55 mol/l (0.55 N) in aqueous solution
Catalog # PRLS89305.440
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pk:20 L
- Seq:5302
Specifications
About this item
:
:40 g/mol
Density:1.04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:3266
ADR:8,III
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548