0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Sodium thiosulphate
Sodium thiosulphate 21% in aqueous solution
Sodium thiosulphate 21% in aqueous solution
PRLSMC312935.9010
:
PROLABO CMP
:
Sodium thiosulphate 21% in aqueous solution
PRLSMC312935.9010
:
PROLABO CMP
:
Pack type:
Plastic container
Pk:
10 L
Seq:
5214
:
:
158.11 g/mol
:
Ambient
:
231-867-5
:
7772-98-7
:
Na₂S₂O₃
:
MFCD00003499