Order Entry
France
ContactUsLinkComponent
Potassium thiocyanate 9.7% in aqueous solution
Potassium thiocyanate 9.7% in aqueous solution
Catalog # PRLSMC306819.0500
Supplier:  PROLABO CMP
CAS Number:  
Potassium thiocyanate 9.7% in aqueous solution
Catalog # PRLSMC306819.0500
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413