Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hypochlorite
Sodium hypochlorite (2 g/l Cl₂) in aqueous solution
Sodium hypochlorite (2 g/l Cl₂) in aqueous solution
Sodium hypochlorite (2 g/l Cl₂) in aqueous solution
PRLS85470.300
:
PROLABO CMP
:
Sodium hypochlorite (2 g/l Cl₂) in aqueous solution
PRLS85470.300
:
PROLABO CMP
:
Pack type:
Plastic bottle
Pk:
2 L
Seq:
5086
:
:
74.44 g/mol
:
Refrigerator
:
231-668-3
:
7681-52-9
:
ClNaO
:
MFCD00011120