Trichloroacetic acid (TCA) 24% in aqueous solution
PRLS88986.360
:
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
: TCA
:
:163.39 g/mol
:196 °C (1013 hPa)
:54…56 °C
:≤1.07 g/cm³ (20 °C)
:Ambient
:200-927-2
:2564
:8,II
:GHS05
:GHS07
:GHS09
:76-03-9
:Cl₃CCOOH
:MFCD00004177