Order Entry
France
ContactUsLinkComponent
Potassium iodide 17% in aqueous solution
Health
Potassium iodide 17% in aqueous solution
Catalog # PRLSMC307203.2000
Supplier:  PROLABO CMP
CAS Number:  
Health
Potassium iodide 17% in aqueous solution
Catalog # PRLSMC307203.2000
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

About this item

 :
 :166 g/mol
EINECS:231-659-4
 :GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405