
Sodium hydroxide 177 g/l in aqueous solution
PRLSMC301000.9010
:
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
:
:40 g/mol
:119…135 °C (1013 hPa)
:9…10 °C
:1.53 g/cm³ (20 °C)
:Ambient
:215-185-5
:1824
:8,II
:GHS05
:1310-73-2
:NaOH
:MFCD00003548