
Sodium hydroxide 540 g/l in aqueous solution
PRLS89289.460
:
- Pack type:Plastic container
- Pk:25 L
- Seq:5327
:
:40 g/mol
:119…135 °C (1013 hPa)
:9…10 °C
:1.53 g/cm³ (20 °C)
:Ambient
:215-185-5
:1824
:8,II
:GHS05
:1310-73-2
:NaOH
:MFCD00003548