
Sodium hydroxide 200 g/l in aqueous solution
PRLS87097.260
:
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
:
:40 g/mol
:119…135 °C (1013 hPa)
:9…10 °C
:1.53 g/cm³ (20 °C)
:Ambient
:215-185-5
:1824
:8,II
:GHS05
:1310-73-2
:NaOH
:MFCD00003548