Order Entry
France
ContactUsLinkComponent
Potassium iodide 0.0125 mol/l (0.0125 N) in aqueous solution
Health
Potassium iodide 0.0125 mol/l (0.0125 N) in aqueous solution
Catalog # PRLS88180.260
Supplier:  PROLABO CMP
CAS Number:  
Health
Potassium iodide 0.0125 mol/l (0.0125 N) in aqueous solution
Catalog # PRLS88180.260
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :166 g/mol
EINECS:231-659-4
 :GHS08
Storage Temperature:Ambient
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405