Order Entry
France
ContactUsLinkComponent
 
Potassium hydroxide 0.01 mol/l (0.01 N) in methanol
Catalog # VWRS5337.1000
Supplier:  VWR Collection
CAS Number:  
Potassium hydroxide 0.01 mol/l (0.01 N) in methanol
Catalog # VWRS5337.1000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Potash caustic, Caustic potash
 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1230
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553