Tubing sleeves, FEP, NanoTight™, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
FEP.
NanoTight tubing sleeves are manufactured using versatile FEP and are precisely cut to a 1,6" length. We offer a wide assortment of sleeves ensuring availability for most applications. Many of the sleeves feature a light colour tint that can help more easily identify the inner diameter for future orders. Because FEP is the base polymer for these sleeves, there is a maximum recommended continuous operating temperature of 50 °C.
Frequently Bought Together