Safety shoes, lace-up, Compositelite™ WORK FC14
Supplier: Portwest
Certificates
About this item
Metal-free, anti-static, lightweight safety shoe made from split leather upper plus an oil resistant, dual-density sole.
- Shock-absorbent heel
- Composite toecap (200 J)
- Puncture resistant composite midsole
Colour: Black.
Certifications: EN 20345:2011 + A1:2007; S1P, SRC.
Frequently Bought Together