Order Entry
France
ContactUsLinkComponent
Disinfectant wipes, Bacillol® AF Tissues
Disinfectant wipes, Bacillol® AF Tissues
  115-2157
undefined
Disinfectant wipes, Bacillol® AF Tissues
  115-2157
  • L×W:
    180×200 mm
  • Pk:
    1
Bacillol® AF Tissues are pre-soaked, ready to use disinfectant wipes made from high quality, tear resistant and absorbent PET fleece. The aldehyde-, colourant- and fragrance-free wipes act rapidly and comprehensively. They are suitable for disinfection of alcohol resistant surfaces, such as work surfaces in medical practices, labs and clinics, and also medical equipment.
  • Can also be used without gloves thanks to the skin friendly formulation
  • Safe and easy dispensing of individual wipes from resealable flowpack
  • Broad range of applications and material compatibility
  • Bacillol® AF saturation fluid guarantees effective moistening
Microbiological effectiveness: bactericidal, yeasticidal, fungicidal, tuberculocidal, mycobactericidal, limited virucidal action (incl. HBV, HIV, HCV), MNV, adeno-, polyoma- and rotaviruses.
VAH, Robert Koch Institute (RKI) list of tested and recognised disinfectants and methods.
 : 80 wipes per resealable, handy flowpack. A wall mounting for the pack is also available.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904