Order Entry
France
ContactUsLinkComponent
D-(+)-Glucose 0.8% in aqueous solution
D-(+)-Glucose 0.8% in aqueous solution
Catalog # PRLS87142.180
Supplier:  PROLABO CMP
CAS Number:  
undefined
D-(+)-Glucose 0.8% in aqueous solution
Catalog # PRLS87142.180
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912