Order Entry
France
ContactUsLinkComponent
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Catalog # MOLEM79047769
Supplier:  Molekula
CAS Number:  
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Catalog # MOLEM79047769
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :520.4 g/mol
CAS Number:128625-52-5
Formula:C₁₈H₂₈F₆N₆OP₂
MDL Number:MFCD00077411