Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Mercury (II) chloride
Mercury(II) chloride 54 g/l in aqueous solution
Mercury(II) chloride 54 g/l in aqueous solution
:
PROLABO CMP
Mercury(II) chloride 54 g/l in aqueous solution
:
PROLABO CMP
:
7487-94-7
:
:
HgCl₂
:
MFCD00011041
:
271.5 g/mol
:
Ambient
:
231-299-8
:
2922
:
8,III
:
GHS06
:
GHS05
:
GHS08
Frequently Bought Together
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
VWR® Rota®, pH indicator paper, 1 to 14
: VWR Chemicals
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Giemsa's stain solution for microscopy, Sigma-Aldrich®
: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Immersion oil for microscopy, Sigma-Aldrich®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
Test strips for peroxide, MQuant®, Supelco®
: Merck
Sodium hydroxide, pellets, pure, Supelco®
: Merck
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Potassium dichromate 106 g/L 0.36 mol/l in aqueous solution Reag. Ph. Eur. 1069501
: VWR Chemicals