
4-Fluorobenzoic anhydride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25569-77-1
:
Formula:C₁₄H₈F₂O₃
MDL Number:MFCD00425938
:262.21 g/mol
Melting Pt:111…115 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together