
3-Methoxydiphenylamine 98%, Sigma-Aldrich®
Catalog # 275980-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to brown powder, crystals, crystalline Powder and/or chunks
- Infrared spectrum:Conforms to structure
- Elemental analysis:Carbon 76.4 - 80.3%, Nitrogen 6.7 - 7.3%
- Gas liquid:97.5% (minimum)
Specification Test Results
About this item
:
:199.25 g/mol
Melting Pt:72...74 °C (lit.)
:M-Clarity™ quality level = MQ100
EINECS:202-921-5
:GHS07
Storage Temperature:Room Temperature
CAS Number:101-16-6
Formula:C₁₃H₁₃NO
MDL Number:MFCD00008383