Order Entry
France
ContactUsLinkComponent
2,3-Dimethylmaleic anhydride for synthesis, Sigma-Aldrich®
Irritant
2,3-Dimethylmaleic anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.41502.0005
Supplier:  Merck
CAS Number:  
Irritant
2,3-Dimethylmaleic anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.41502.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Melting Point:
    96 °C
  • Molar Mass:
    126.11 g/mol
  • Boiling Point:
    223 °C
  • Density:
    1.107 g/cm3 (100 °C)

Specification Test Results

About this item

 :
 :126.11 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:212-165-8
 :GHS07
Storage Temperature:Ambient
CAS Number:766-39-2
Formula:C₆H₆O₃
MDL Number:MFCD00005523