Order Entry
France
ContactUsLinkComponent
N-Methylisatoic anhydride tech. 90%, Technical Grade
N-Methylisatoic anhydride tech. 90%, Technical Grade
Catalog # B24099.22
CAS Number:  
N-Methylisatoic anhydride tech. 90%, Technical Grade
Catalog # B24099.22
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :177.16 g/mol
Melting Pt:∼ 160 °C (dec.)
EINECS:233-714-8
 :GHS07
Storage Temperature:Ambient
CAS Number:10328-92-4
Formula:C₉H₇NO₃
MDL Number:MFCD00006815